Assay Details
| Gene Symbol: Su(z)12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143802.3 | NP_652059.1 | 4 - 5 | 2053 | 61 | |
| NM_168826.2 | NP_730465.1 | 4 - 5 | 2053 | 61 | ||
| GenBank mRNA | AF149047.1 | - | 4 - 5 | 1798 | 61 | |
| AY069809.1 | - | 4 - 5 | 1991 | 61 | ||
More Information
Set Membership: |
|

Su(z)12
Genomic Map