Assay Details
| Gene Symbol: Gabat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170008.1 | NP_001163479.1 | 2 - 3 | 318 | 72 | |
| NM_001300196.1 | NP_001287125.1 | 1 - 2 | 124 | 72 | ||
| NM_140911.4 | NP_649168.2 | 1 - 2 | 239 | 72 | ||
| NM_168829.3 | NP_730476.1 | 3 - 4 | 190 | 72 | ||
More Information
Set Membership: |
|

Gabat
Genomic Map