Assay Details
| Gene Symbol: Gabat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170008.1 | NP_001163479.1 | 5 - 6 | 775 | 68 | |
| NM_001300196.1 | NP_001287125.1 | 4 - 5 | 581 | 68 | ||
| NM_140911.4 | NP_649168.2 | 4 - 5 | 696 | 68 | ||
| NM_168829.3 | NP_730476.1 | 6 - 7 | 647 | 68 | ||
More Information
Set Membership: |
|

Gabat
Genomic Map