Assay Details
| Gene Symbol: eRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275175.1 | NP_001262104.1 | 5 - 6 | 1095 | 73 | |
| NM_001275176.1 | NP_001262105.1 | 5 - 6 | 1095 | 73 | ||
| NM_140953.3 | NP_649210.1 | 4 - 5 | 1651 | 73 | ||
| NM_168845.2 | NP_730517.1 | 5 - 6 | 1131 | 73 | ||
| NM_168846.2 | NP_730518.1 | 5 - 6 | 1095 | 73 | ||
| NM_168847.2 | NP_730519.1 | 5 - 6 | 1360 | 73 | ||
| NM_168848.2 | NP_730520.1 | 5 - 6 | 1118 | 73 | ||
| NM_176369.3 | NP_788547.1 | 4 - 5 | 1069 | 73 | ||
| GenBank mRNA | AY069511.1 | - | 4 - 5 | 1096 | 73 | |
| BT120009.1 | - | 5 - 6 | 1102 | 73 | ||
| BT120010.1 | - | 5 - 6 | 1105 | 73 | ||
More Information
Set Membership: |
|

eRF1
Genomic Map