Assay Details
| Gene Symbol: Pka-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014593.3 | NP_001014593.1 | 1 - 2 | 712 | 107 | |
| NM_001014594.3 | NP_001014594.1 | 1 - 2 | 712 | 107 | ||
| NM_001014595.3 | NP_001014595.1 | 2 - 3 | 647 | 107 | ||
| NM_001014596.3 | NP_001014596.1 | 2 - 3 | 317 | 107 | ||
| NM_001202218.2 | NP_001189147.1 | 1 - 2 | 712 | 107 | ||
| NM_001202219.2 | NP_001189148.1 | 2 - 3 | 654 | 107 | ||
| NM_001202220.2 | NP_001189149.1 | 2 - 3 | 317 | 107 | ||
| NM_168875.4 | NP_730573.2 | 1 - 2 | 712 | 107 | ||
| NM_168876.6 | NP_730574.2 | 2 - 3 | 654 | 107 | ||
| GenBank mRNA | AY058709.1 | - | 1 - 2 | 678 | 107 | |
More Information
Set Membership: |
|

Pka-R1
Genomic Map