Assay Details
| Gene Symbol: Pka-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014593.3 | NP_001014593.1 | 4 - 5 | 1586 | 59 | |
| NM_001014594.3 | NP_001014594.1 | 4 - 5 | 1586 | 59 | ||
| NM_001014595.3 | NP_001014595.1 | 5 - 6 | 1521 | 59 | ||
| NM_001014596.3 | NP_001014596.1 | 5 - 6 | 1191 | 59 | ||
| NM_001014597.2 | NP_001014597.1 | 4 - 5 | 1019 | 59 | ||
| NM_001014598.2 | NP_001014598.1 | 4 - 5 | 1266 | 59 | ||
| NM_001202215.1 | NP_001189144.1 | 5 - 6 | 1145 | 59 | ||
| NM_001202216.1 | NP_001189145.1 | 4 - 5 | 1196 | 59 | ||
| NM_001202217.1 | NP_001189146.1 | 4 - 5 | 1269 | 59 | ||
| NM_001202218.2 | NP_001189147.1 | 4 - 5 | 1589 | 59 | ||
| NM_001202219.2 | NP_001189148.1 | 5 - 6 | 1531 | 59 | ||
| NM_001202220.2 | NP_001189149.1 | 5 - 6 | 1194 | 59 | ||
| NM_001202221.1 | NP_001189150.1 | 4 - 5 | 1378 | 59 | ||
| NM_079465.4 | NP_524189.2 | 4 - 5 | 1193 | 59 | ||
| NM_141006.4 | NP_649263.2 | 4 - 5 | 1375 | 59 | ||
| NM_168875.4 | NP_730573.2 | 4 - 5 | 1586 | 59 | ||
| NM_168876.6 | NP_730574.2 | 5 - 6 | 1528 | 59 | ||
| NM_168877.2 | NP_730576.1 | 5 - 6 | 1142 | 59 | ||
| GenBank mRNA | AY058709.1 | - | 4 - 5 | 1552 | 59 | |
| AY122131.1 | - | 4 - 5 | 1193 | 59 | ||
| BT133037.1 | - | 5 - 6 | 1145 | 59 | ||
More Information
Set Membership: |
|

Pka-R1
Genomic Map