Assay Details
| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275205.1 | NP_001262134.1 | 10 - 11 | 7799 | 67 | |
| NM_079914.3 | NP_524653.1 | 10 - 11 | 8282 | 67 | ||
| NM_168879.3 | NP_730590.1 | 10 - 11 | 7914 | 67 | ||
| NM_168880.3 | NP_730591.2 | 11 - 12 | 8520 | 67 | ||
| GenBank mRNA | AF226855.1 | - | 10 - 11 | 8274 | 67 | |
| AF227214.1 | - | 9 - 10 | 7488 | 67 | ||
| AF324425.1 | - | 10 - 11 | 7832 | 67 | ||
| BT125901.1 | - | 10 - 11 | 7832 | 67 | ||
More Information
Set Membership: |
|

skd
Genomic Map