Assay Details
| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275205.1 | NP_001262134.1 | 11 - 12 | 7907 | 67 | |
| NM_079914.3 | NP_524653.1 | 11 - 12 | 8390 | 67 | ||
| NM_168879.3 | NP_730590.1 | 11 - 12 | 8022 | 67 | ||
| NM_168880.3 | NP_730591.2 | 12 - 13 | 8628 | 67 | ||
| GenBank mRNA | AF226855.1 | - | 11 - 12 | 8382 | 67 | |
| AF227214.1 | - | 10 - 11 | 7596 | 67 | ||
| AF324425.1 | - | 11 - 12 | 7940 | 67 | ||
| BT125901.1 | - | 11 - 12 | 7940 | 67 | ||
More Information
Set Membership: |
|

skd
Genomic Map