Assay Details
| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275205.1 | NP_001262134.1 | 6 - 7 | 1609 | 87 | |
| NM_079914.3 | NP_524653.1 | 6 - 7 | 2041 | 87 | ||
| NM_168879.3 | NP_730590.1 | 6 - 7 | 1673 | 87 | ||
| NM_168880.3 | NP_730591.2 | 6 - 7 | 1829 | 87 | ||
| GenBank mRNA | AF226855.1 | - | 6 - 7 | 2033 | 87 | |
| AF227214.1 | - | 5 - 6 | 1247 | 87 | ||
| AF324425.1 | - | 6 - 7 | 1591 | 87 | ||
| BT125901.1 | - | 6 - 7 | 1591 | 87 | ||
More Information
Set Membership: |
|

skd
Genomic Map