Assay Details
| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275205.1 | NP_001262134.1 | 7 - 8 | 2300 | 74 | |
| NM_079914.3 | NP_524653.1 | 7 - 8 | 2732 | 74 | ||
| NM_168879.3 | NP_730590.1 | 7 - 8 | 2364 | 74 | ||
| GenBank mRNA | AF226855.1 | - | 7 - 8 | 2724 | 74 | |
| AF227214.1 | - | 6 - 7 | 1938 | 74 | ||
| AF324425.1 | - | 7 - 8 | 2282 | 74 | ||
| BT125901.1 | - | 7 - 8 | 2282 | 74 | ||
More Information
Set Membership: |
|

skd
Genomic Map