Assay Details
| Gene Symbol: skd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079914.3 | NP_524653.1 | 8 - 9 | 2911 | 65 | |
| NM_168879.3 | NP_730590.1 | 8 - 9 | 2543 | 65 | ||
| NM_168880.3 | NP_730591.2 | 9 - 10 | 3149 | 65 | ||
| GenBank mRNA | AF226855.1 | - | 8 - 9 | 2903 | 65 | |
| AF227214.1 | - | 7 - 8 | 2117 | 65 | ||
| AF324425.1 | - | 8 - 9 | 2461 | 65 | ||
| BT125901.1 | - | 8 - 9 | 2461 | 65 | ||
More Information
Set Membership: |
|

skd
Genomic Map