Assay Details
| Gene Symbol: Nak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299146.1 | NP_001286075.1 | 2 - 3 | 627 | 76 | |
| NM_057817.4 | NP_477165.1 | 2 - 3 | 627 | 76 | ||
| NM_057818.4 | NP_477166.2 | 2 - 3 | 627 | 76 | ||
| NM_206003.2 | NP_995725.1 | 2 - 3 | 604 | 76 | ||
More Information
Set Membership: |
|

Nak
Genomic Map