Assay Details
| Gene Symbol: Nak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299146.1 | NP_001286075.1 | 3 - 4 | 758 | 73 | |
| NM_057817.4 | NP_477165.1 | 3 - 4 | 758 | 73 | ||
| NM_057818.4 | NP_477166.2 | 3 - 4 | 758 | 73 | ||
| NM_206003.2 | NP_995725.1 | 3 - 4 | 735 | 73 | ||
More Information
Set Membership: |
|

Nak
Genomic Map