Assay Details
| Gene Symbol: jing | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103729.2 | NP_001097199.1 | 2 - 3 | 2048 | 67 | |
| NM_001103730.2 | NP_001097200.1 | 2 - 3 | 3319 | 67 | ||
| NM_136371.3 | NP_610215.3 | 2 - 3 | 2048 | 67 | ||
| NM_206027.3 | NP_995749.2 | 2 - 3 | 1739 | 67 | ||
| GenBank mRNA | AF285778.1 | - | 1 - 2 | 1512 | 67 | |
| BT025201.1 | - | 2 - 3 | 1719 | 67 | ||
More Information
Set Membership: |
|

jing
Genomic Map