Assay Details
| Gene Symbol: Spn42Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165495.4 | NP_724511.2 | 2 - 3 | 1646 | 90 | |
| NM_206032.2 | NP_995754.1 | 3 - 4 | 1280 | 90 | ||
| NM_206035.2 | NP_995757.1 | 3 - 4 | 1280 | 90 | ||
| NM_206037.3 | NP_995759.1 | 3 - 4 | 1160 | 90 | ||
| NM_206038.3 | NP_995760.1 | 3 - 4 | 1160 | 90 | ||
| GenBank mRNA | AJ428881.1 | - | 3 - 4 | 1154 | 90 | |
| AJ428882.1 | - | 3 - 4 | 1154 | 90 | ||
| AJ428886.1 | - | 3 - 4 | 1280 | 90 | ||
| AJ428887.1 | - | 3 - 4 | 1280 | 90 | ||
| BT001604.1 | - | 3 - 4 | 1280 | 90 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Spn42Da
Spn42Da Genomic Map
Genomic Map Back To Top
 Back To Top