Assay Details
| Gene Symbol: Spn42Da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165495.4 | NP_724511.2 | 3 - 4 | 1777 | 118 | |
| NM_206032.2 | NP_995754.1 | 4 - 5 | 1411 | 118 | ||
| NM_206037.3 | NP_995759.1 | 4 - 5 | 1291 | 118 | ||
| GenBank mRNA | AJ428881.1 | - | 4 - 5 | 1285 | 118 | |
| AJ428886.1 | - | 4 - 5 | 1411 | 118 | ||
| BT001604.1 | - | 4 - 5 | 1411 | 118 | ||
More Information
Set Membership: |
|

Spn42Da
Genomic Map