Assay Details
| Gene Symbol: mim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014500.3 | NP_001014500.2 | 5 - 6 | 1551 | 67 | |
| NM_001103735.3 | NP_001097205.1 | 5 - 6 | 1551 | 67 | ||
| NM_001169597.2 | NP_001163068.1 | 5 - 6 | 1551 | 67 | ||
| NM_001201939.3 | NP_001188868.1 | 5 - 6 | 1551 | 67 | ||
| NM_001201940.3 | NP_001188869.1 | 5 - 6 | 1551 | 67 | ||
| NM_001201941.2 | NP_001188870.1 | 5 - 6 | 1551 | 67 | ||
| NM_001259228.2 | NP_001246157.1 | 5 - 6 | 1551 | 67 | ||
| NM_001259229.2 | NP_001246158.1 | 5 - 6 | 1551 | 67 | ||
| NM_001259230.2 | NP_001246159.1 | 5 - 6 | 1551 | 67 | ||
| NM_001273818.1 | NP_001260747.1 | 5 - 6 | 1551 | 67 | ||
| NM_001273819.1 | NP_001260748.1 | 5 - 6 | 1551 | 67 | ||
| NM_001273820.1 | NP_001260749.1 | 5 - 6 | 1551 | 67 | ||
| NM_001273821.1 | NP_001260750.1 | 5 - 6 | 1551 | 67 | ||
More Information
Set Membership: |
|

mim
Genomic Map