Assay Details
| Gene Symbol: mim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014500.3 | NP_001014500.2 | 10 - 11 | 4663 | 105 | |
| NM_001103735.3 | NP_001097205.1 | 10 - 11 | 4663 | 105 | ||
| NM_001201940.3 | NP_001188869.1 | 10 - 11 | 4663 | 105 | ||
| NM_001201941.2 | NP_001188870.1 | 10 - 11 | 4609 | 105 | ||
| NM_001259229.2 | NP_001246158.1 | 11 - 12 | 4744 | 105 | ||
| NM_001259230.2 | NP_001246159.1 | 11 - 12 | 4744 | 105 | ||
| NM_001273819.1 | NP_001260748.1 | 11 - 12 | 4258 | 105 | ||
| NM_001273821.1 | NP_001260750.1 | 11 - 12 | 4744 | 105 | ||
More Information
Set Membership: |
|

mim
Genomic Map