Assay Details
| Gene Symbol: Eaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136429.4 | NP_610273.1 | 2 - 3 | 937 | 83 | |
| NM_165518.3 | NP_724547.1 | 2 - 3 | 1539 | 83 | ||
| NM_165519.3 | NP_724548.1 | 1 - 2 | 1616 | 83 | ||
| GenBank mRNA | BT001503.1 | - | 1 - 2 | 1592 | 83 | |
| BT001556.1 | - | 2 - 3 | 926 | 83 | ||
More Information
Set Membership: |
|

Eaf
Genomic Map