Assay Details
| Gene Symbol: Eaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136429.4 | NP_610273.1 | 4 - 5 | 1165 | 81 | |
| NM_165518.3 | NP_724547.1 | 4 - 5 | 1767 | 81 | ||
| NM_165519.3 | NP_724548.1 | 3 - 4 | 1844 | 81 | ||
| NM_165521.3 | NP_724550.1 | 4 - 5 | 713 | 81 | ||
| GenBank mRNA | BT001503.1 | - | 3 - 4 | 1820 | 81 | |
| BT001556.1 | - | 4 - 5 | 1154 | 81 | ||
| BT003263.1 | - | 4 - 5 | 656 | 81 | ||
More Information
Set Membership: |
|

Eaf
Genomic Map