Assay Details
| Gene Symbol: Eaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136429.4 | NP_610273.1 | 6 - 7 | 1459 | 87 | |
| NM_165518.3 | NP_724547.1 | 6 - 7 | 2061 | 87 | ||
| NM_165519.3 | NP_724548.1 | 5 - 6 | 2138 | 87 | ||
| NM_165521.3 | NP_724550.1 | 6 - 7 | 1007 | 87 | ||
| GenBank mRNA | BT001503.1 | - | 5 - 6 | 2114 | 87 | |
| BT001556.1 | - | 6 - 7 | 1448 | 87 | ||
| BT003263.1 | - | 6 - 7 | 950 | 87 | ||
More Information
Set Membership: |
|

Eaf
Genomic Map