Assay Details
| Gene Symbol: mtt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144147.2 | NP_001137619.1 | 18 - 19 | 5340 | 78 | |
| NM_001273877.1 | NP_001260806.1 | 18 - 19 | 5340 | 78 | ||
| NM_136542.3 | NP_610386.2 | 17 - 18 | 4380 | 78 | ||
| NM_165609.3 | NP_724683.2 | 16 - 17 | 3434 | 78 | ||
| NM_206058.2 | NP_995780.1 | 12 - 13 | 1906 | 78 | ||
| GenBank mRNA | BT033068.1 | - | 12 - 13 | 1907 | 78 | |
More Information
Set Membership: |
|

mtt
Genomic Map