Assay Details
| Gene Symbol: mtt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144147.2 | NP_001137619.1 | 15 - 16 | 4780 | 69 | |
| NM_001273877.1 | NP_001260806.1 | 15 - 16 | 4780 | 69 | ||
| NM_136542.3 | NP_610386.2 | 14 - 15 | 3820 | 69 | ||
| NM_165609.3 | NP_724683.2 | 13 - 14 | 2874 | 69 | ||
| NM_206058.2 | NP_995780.1 | 9 - 10 | 1346 | 69 | ||
| GenBank mRNA | BT033068.1 | - | 9 - 10 | 1347 | 69 | |
More Information
Set Membership: |
|

mtt
Genomic Map