Assay Details
| Gene Symbol: psq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014518.3 | NP_001014518.1 | 6 - 7 | 2927 | 70 | |
| NM_001014519.3 | NP_001014519.1 | 6 - 7 | 3176 | 70 | ||
| NM_001382050.1 | NP_001369079.1 | 7 - 8 | 3012 | 70 | ||
| NM_001382051.1 | NP_001369080.1 | 7 - 8 | 3012 | 70 | ||
| NM_078962.4 | NP_523686.2 | 9 - 10 | 3654 | 70 | ||
| NM_165790.3 | NP_724955.1 | 10 - 11 | 3600 | 70 | ||
| NM_165791.3 | NP_724958.1 | 7 - 8 | 3012 | 70 | ||
| NM_165792.3 | NP_724959.1 | 8 - 9 | 2751 | 70 | ||
| NM_176141.3 | NP_788321.1 | 8 - 9 | 3067 | 70 | ||
| NM_176142.3 | NP_788322.1 | 8 - 9 | 2800 | 70 | ||
| NM_176143.3 | NP_788323.1 | 7 - 8 | 2837 | 70 | ||
| NM_206086.3 | NP_995808.1 | 9 - 10 | 3497 | 70 | ||
| GenBank mRNA | AY069588.1 | - | 9 - 10 | 3575 | 70 | |
More Information
Set Membership: |
|

psq
Genomic Map