Assay Details
| Gene Symbol: shn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273953.1 | NP_001260882.1 | 3 - 4 | 1797 | 60 | |
| NM_001273954.1 | NP_001260883.1 | 3 - 4 | 2672 | 60 | ||
| NM_057375.4 | NP_476723.1 | 3 - 4 | 1726 | 60 | ||
| NM_057376.3 | NP_476724.2 | 2 - 3 | 1812 | 60 | ||
| NM_134279.3 | NP_599106.1 | 2 - 3 | 2338 | 60 | ||
| NM_206091.2 | NP_995813.1 | 3 - 4 | 2672 | 60 | ||
More Information
Set Membership: |
|

shn
Genomic Map