Assay Details
| Gene Symbol: Cap-G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169664.2 | NP_001163135.1 | 1 - 2 | 423 | 65 | |
| NM_206105.4 | NP_995827.2 | 1 - 2 | 423 | 65 | ||
| GenBank mRNA | BT032841.1 | - | 1 - 2 | 363 | 65 | |
More Information
Set Membership: |
|

Cap-G
Genomic Map