Assay Details
| Gene Symbol: Fs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299516.1 | NP_001286445.1 | 2 - 3 | 1040 | 102 | |
| NM_144119.3 | NP_652376.2 | 3 - 4 | 1308 | 102 | ||
| GenBank mRNA | BT003621.1 | - | 3 - 4 | 1298 | 102 | |
More Information
Set Membership: |
|

Fs
Genomic Map