Assay Details
| Gene Symbol: Fs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299516.1 | NP_001286445.1 | 3 - 4 | 1226 | 76 | |
| NM_144119.3 | NP_652376.2 | 4 - 5 | 1494 | 76 | ||
| GenBank mRNA | BT003621.1 | - | 4 - 5 | 1484 | 76 | |
More Information
Set Membership: |
|

Fs
Genomic Map