Assay Details
| Gene Symbol: Fs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299516.1 | NP_001286445.1 | 7 - 8 | 2595 | 58 | |
| NM_144119.3 | NP_652376.2 | 8 - 9 | 2866 | 58 | ||
| GenBank mRNA | BT003621.1 | - | 8 - 9 | 2856 | 58 | |
More Information
Set Membership: |
|

Fs
Genomic Map