Assay Details
| Gene Symbol: casp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299544.1 | NP_001286473.1 | 11 - 12 | 1919 | 58 | |
| NM_137236.5 | NP_611080.1 | 11 - 12 | 1934 | 58 | ||
| NM_206126.2 | NP_995848.1 | 12 - 13 | 1934 | 58 | ||
| GenBank mRNA | AY122148.1 | - | 11 - 12 | 1875 | 58 | |
More Information
Set Membership: |
|

casp
Genomic Map