Assay Details
| Gene Symbol: Rho1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057750.4 | NP_477098.1 | 2 - 3 | 431 | 56 | |
| NM_134308.3 | NP_599135.1 | 2 - 3 | 431 | 56 | ||
| NM_134309.2 | NP_599136.1 | 2 - 3 | 302 | 56 | ||
| NM_166139.3 | NP_725524.1 | 1 - 2 | 313 | 56 | ||
| NM_206127.2 | NP_995849.1 | 2 - 3 | 380 | 56 | ||
| NM_206128.2 | NP_995850.1 | 2 - 3 | 374 | 56 | ||
| GenBank mRNA | AF177872.1 | - | 2 - 3 | 384 | 56 | |
| AF177873.1 | - | 2 - 3 | 339 | 56 | ||
| AF177874.2 | - | 2 - 3 | 201 | 56 | ||
| AY119536.1 | - | 1 - 2 | 297 | 56 | ||
| BT010085.1 | - | 2 - 3 | 373 | 56 | ||
| L38311.1 | - | 201 | 56 | |||
More Information
Set Membership: |
|

Rho1
Genomic Map