Assay Details
| Gene Symbol: RhoGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299586.1 | NP_001286515.1 | 15 - 16 | 4210 | 79 | |
| NM_057969.6 | NP_477317.1 | 15 - 16 | 4519 | 79 | ||
| NM_206146.4 | NP_995868.1 | 15 - 16 | 4530 | 79 | ||
| NM_206147.4 | NP_995869.1 | 14 - 15 | 3468 | 79 | ||
More Information
Set Membership: |
|

RhoGEF2
Genomic Map