Assay Details
| Gene Symbol: RhoGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259452.2 | NP_001246381.1 | 18 - 19 | 5698 | 66 | |
| NM_001299586.1 | NP_001286515.1 | 18 - 19 | 5384 | 66 | ||
| NM_057969.6 | NP_477317.1 | 18 - 19 | 5693 | 66 | ||
| NM_206146.4 | NP_995868.1 | 18 - 19 | 5704 | 66 | ||
| NM_206147.4 | NP_995869.1 | 17 - 18 | 4642 | 66 | ||
More Information
Set Membership: |
|

RhoGEF2
Genomic Map