Assay Details
| Gene Symbol: RhoGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259452.2 | NP_001246381.1 | 7 - 8 | 1758 | 61 | |
| NM_057969.6 | NP_477317.1 | 7 - 8 | 1747 | 61 | ||
| NM_206146.4 | NP_995868.1 | 7 - 8 | 1758 | 61 | ||
More Information
Set Membership: |
|

RhoGEF2
Genomic Map