Assay Details
| Gene Symbol: EDTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079046.3 | NP_523770.2 | 4 - 5 | 2236 | 81 | |
| NM_206151.2 | NP_995873.2 | 4 - 5 | 2257 | 81 | ||
| GenBank mRNA | AB036800.2 | - | 4 - 5 | 2230 | 81 | |
| AF145681.1 | - | 4 - 5 | 2214 | 81 | ||
More Information
Set Membership: |
|

EDTP
Genomic Map