Assay Details
| Gene Symbol: Patronin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144218.3 | NP_001137690.1 | 10 - 11 | 1914 | 65 | |
| NM_001144219.3 | NP_001137691.1 | 11 - 12 | 1977 | 65 | ||
| NM_001169704.2 | NP_001163175.1 | 11 - 12 | 1977 | 65 | ||
| NM_001169706.2 | NP_001163177.2 | 11 - 12 | 1977 | 65 | ||
| NM_001274122.1 | NP_001261051.1 | 11 - 12 | 1977 | 65 | ||
| NM_001274123.1 | NP_001261052.1 | 12 - 13 | 2130 | 65 | ||
| NM_001274124.1 | NP_001261053.1 | 11 - 12 | 2025 | 65 | ||
| NM_001274125.1 | NP_001261054.1 | 10 - 11 | 1872 | 65 | ||
| NM_001274126.1 | NP_001261055.1 | 9 - 10 | 1731 | 65 | ||
| NM_001274127.1 | NP_001261056.1 | 8 - 9 | 1563 | 65 | ||
| NM_176218.3 | NP_788398.1 | 10 - 11 | 2392 | 65 | ||
| NM_206153.3 | NP_995875.1 | 10 - 11 | 1914 | 65 | ||
| NM_206154.3 | NP_995876.1 | 11 - 12 | 1977 | 65 | ||
More Information
Set Membership: |
|

Patronin
Genomic Map