Assay Details
| Gene Symbol: olf186-F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144224.2 | NP_001137696.1 | 1 - 2 | 2561 | 62 | |
| NM_001274134.1 | NP_001261063.1 | 2 - 3 | 3264 | 62 | ||
| NM_137429.3 | NP_611273.1 | 4 - 5 | 1314 | 62 | ||
| NM_170626.2 | NP_725727.1 | 2 - 3 | 762 | 62 | ||
| NM_206159.2 | NP_995881.2 | 3 - 4 | 1089 | 62 | ||
| GenBank mRNA | AF188634.1 | - | 4 - 5 | 836 | 62 | |
| AY071273.1 | - | 4 - 5 | 1312 | 62 | ||
| BT001876.1 | - | 1 - 2 | 2295 | 62 | ||
More Information
Set Membership: |
|

olf186-F
Genomic Map