Assay Details
| Gene Symbol: pAbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057319.5 | NP_476667.1 | 2 - 3 | 367 | 80 | |
| NM_166264.4 | NP_725749.1 | 2 - 3 | 231 | 80 | ||
| NM_166265.2 | NP_725750.1 | 2 - 3 | 444 | 80 | ||
| GenBank mRNA | AY118528.1 | - | 2 - 3 | 354 | 80 | |
More Information
Set Membership: |
|

pAbp
Genomic Map