Assay Details
| Gene Symbol: pAbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057319.5 | NP_476667.1 | 3 - 4 | 1845 | 66 | |
| NM_166264.4 | NP_725749.1 | 3 - 4 | 1709 | 66 | ||
| NM_166265.2 | NP_725750.1 | 3 - 4 | 1922 | 66 | ||
| GenBank mRNA | AY118528.1 | - | 3 - 4 | 1832 | 66 | |
More Information
Set Membership: |
|

pAbp
Genomic Map