Assay Details
| Gene Symbol: Dp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299644.1 | NP_001286573.1 | 4 - 5 | 2732 | 65 | |
| NM_079057.3 | NP_523781.2 | 4 - 5 | 2377 | 65 | ||
| NM_166280.2 | NP_725785.1 | 5 - 6 | 2601 | 65 | ||
| NM_166281.2 | NP_725786.1 | 5 - 6 | 2374 | 65 | ||
| NM_166282.2 | NP_725787.1 | 5 - 6 | 2529 | 65 | ||
| NM_206163.2 | NP_995885.1 | 5 - 6 | 2301 | 65 | ||
| NM_206164.2 | NP_995886.1 | 5 - 6 | 2298 | 65 | ||
| GenBank mRNA | AF132179.1 | - | 4 - 5 | 2271 | 65 | |
More Information
Set Membership: |
|

Dp1
Genomic Map