Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014540.3 | NP_001014540.2 | 12 - 13 | 3306 | 68 | |
| NM_001014542.3 | NP_001014542.1 | 12 - 13 | 3309 | 68 | ||
| NM_001169738.2 | NP_001163209.1 | 12 - 13 | 3361 | 68 | ||
| NM_001169739.2 | NP_001163210.1 | 12 - 13 | 3291 | 68 | ||
| NM_001169740.2 | NP_001163211.1 | 12 - 13 | 3441 | 68 | ||
| NM_206171.4 | NP_995893.1 | 13 - 14 | 3319 | 68 | ||
| NM_206172.3 | NP_995894.2 | 12 - 13 | 3309 | 68 | ||
| NM_206178.3 | NP_995900.1 | 12 - 13 | 3309 | 68 | ||
| GenBank mRNA | AF258462.1 | - | 12 - 13 | 3284 | 68 | |
| AF387635.1 | - | 12 - 13 | 3281 | 68 | ||
| AF387636.1 | - | 11 - 12 | 3217 | 68 | ||
| BT009940.1 | - | 12 - 13 | 3313 | 68 | ||
| BT011020.1 | - | 12 - 13 | 3304 | 68 | ||
More Information
Set Membership: |
|

par-1
Genomic Map