Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014540.3 | NP_001014540.2 | 13 - 14 | 3431 | 114 | |
| NM_001014542.3 | NP_001014542.1 | 13 - 14 | 3434 | 114 | ||
| NM_001169738.2 | NP_001163209.1 | 13 - 14 | 3486 | 114 | ||
| NM_001169739.2 | NP_001163210.1 | 13 - 14 | 3416 | 114 | ||
| NM_001169740.2 | NP_001163211.1 | 13 - 14 | 3566 | 114 | ||
| NM_206171.4 | NP_995893.1 | 14 - 15 | 3444 | 114 | ||
| GenBank mRNA | AF387635.1 | - | 13 - 14 | 3406 | 114 | |
| AF387636.1 | - | 12 - 13 | 3342 | 114 | ||
| BT009940.1 | - | 13 - 14 | 3438 | 114 | ||
| BT011020.1 | - | 13 - 14 | 3429 | 114 | ||
More Information
Set Membership: |
|

par-1
Genomic Map