Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014540.3 | NP_001014540.2 | 9 - 10 | 2944 | 95 | |
| NM_001014542.3 | NP_001014542.1 | 9 - 10 | 2929 | 95 | ||
| NM_001169737.2 | NP_001163208.1 | 9 - 10 | 3050 | 95 | ||
| NM_001169738.2 | NP_001163209.1 | 9 - 10 | 3026 | 95 | ||
| NM_001169739.2 | NP_001163210.1 | 9 - 10 | 2929 | 95 | ||
| NM_001169740.2 | NP_001163211.1 | 9 - 10 | 3079 | 95 | ||
| NM_206171.4 | NP_995893.1 | 9 - 10 | 2843 | 95 | ||
| NM_206172.3 | NP_995894.2 | 9 - 10 | 2929 | 95 | ||
| NM_206177.3 | NP_995899.2 | 9 - 10 | 2953 | 95 | ||
| NM_206178.3 | NP_995900.1 | 9 - 10 | 2929 | 95 | ||
| GenBank mRNA | AF258462.1 | - | 9 - 10 | 2904 | 95 | |
| AF387635.1 | - | 9 - 10 | 2919 | 95 | ||
| AF387636.1 | - | 8 - 9 | 2855 | 95 | ||
| BT009940.1 | - | 9 - 10 | 2978 | 95 | ||
| BT011020.1 | - | 9 - 10 | 2924 | 95 | ||
More Information
Set Membership: |
|

par-1
Genomic Map