Assay Details
| Gene Symbol: HnRNP-K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169745.2 | NP_001163216.1 | 6 - 7 | 574 | 77 | |
| NM_001274173.1 | NP_001261102.1 | 6 - 7 | 576 | 77 | ||
| NM_145883.3 | NP_665890.2 | 5 - 6 | 530 | 77 | ||
| NM_206184.2 | NP_995906.1 | 7 - 8 | 625 | 77 | ||
| NM_206185.2 | NP_995907.1 | 6 - 7 | 574 | 77 | ||
| GenBank mRNA | BT032853.1 | - | 6 - 7 | 559 | 77 | |
| BT100100.1 | - | 7 - 8 | 601 | 77 | ||
More Information
Set Membership: |
|

HnRNP-K
Genomic Map