Assay Details
| Gene Symbol: HnRNP-K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274173.1 | NP_001261102.1 | 8 - 9 | 1668 | 82 | |
| NM_145883.3 | NP_665890.2 | 7 - 8 | 1622 | 82 | ||
| NM_206184.2 | NP_995906.1 | 9 - 10 | 1717 | 82 | ||
| NM_206185.2 | NP_995907.1 | 8 - 9 | 1666 | 82 | ||
| GenBank mRNA | BT032853.1 | - | 8 - 9 | 1651 | 82 | |
| BT100100.1 | - | 9 - 10 | 1693 | 82 | ||
More Information
Set Membership: |
|

HnRNP-K
Genomic Map