Assay Details
| Gene Symbol: Slik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166669.4 | NP_726441.1 | 12 - 13 | 5245 | 55 | |
| NM_206213.3 | NP_995935.1 | 12 - 13 | 5638 | 55 | ||
More Information
Set Membership: |
|

Slik
Genomic Map