Assay Details
| Gene Symbol: Slik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014549.3 | NP_001014549.1 | 4 - 5 | 907 | 83 | |
| NM_001144280.2 | NP_001137752.1 | 4 - 5 | 866 | 83 | ||
| NM_138064.4 | NP_611908.2 | 4 - 5 | 907 | 83 | ||
| NM_166669.4 | NP_726441.1 | 4 - 5 | 907 | 83 | ||
| NM_206213.3 | NP_995935.1 | 4 - 5 | 1300 | 83 | ||
| NM_206214.4 | NP_995936.2 | 4 - 5 | 907 | 83 | ||
| GenBank mRNA | BT150436.1 | - | 4 - 5 | 915 | 83 | |
More Information
Set Membership: |
|

Slik
Genomic Map