Assay Details
| Gene Symbol: PIG-Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299911.1 | NP_001286840.1 | 1 - 2 | 253 | 76 | |
| GenBank mRNA | AY118320.1 | - | 1 - 2 | 232 | 76 | |
| Gene Symbol: CG45069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299913.1 | NP_001286842.1 | 1 - 2 | 253 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001299913.1 transcripts.
Set Membership: |
|

PIG-Z
Genomic Map