Assay Details
| Gene Symbol: CG6845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144387.3 | NP_001137859.1 | 2 - 3 | 903 | 154 | |
| NM_138156.5 | NP_612000.3 | 3 - 4 | 848 | 154 | ||
| GenBank mRNA | AY051619.1 | - | 1 - 2 | 263 | 154 | |
| BT003264.1 | - | 2 - 3 | 883 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY051619.1, BT003264.1, NM_001144387.3 transcripts.
Set Membership: |
|

CG6845
Genomic Map