Assay Details
| Gene Symbol: CG6845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144387.3 | NP_001137859.1 | 3 - 4 | 1334 | 70 | |
| NM_138156.5 | NP_612000.3 | 5 - 6 | 1216 | 70 | ||
| GenBank mRNA | AY051619.1 | - | 3 - 4 | 611 | 70 | |
| BT003264.1 | - | 3 - 4 | 1314 | 70 | ||
More Information
Set Membership: |
|

CG6845
Genomic Map